2,3-二溴丙酰氯 ≥98.0% (通过滴定分析)
供应商: TCI
|
Danger
|
D3288-25G
D3288-5G
TCIAD3288-25GEA
0
CNY
TCIAD3288-25G
TCIAD3288-5G
2,3-二溴丙酰氯 ≥98.0% (通过滴定分析)
2,3-二溴丙酰氯
分子式:
C₃H₃Br₂ClO 沸点: 193°C 闪点: 66°C |
MDL编号:
MFCD00000712 CAS号: 18791-02-1 EINECS: 242-575-2 UN号: 3265 ADR: 8,II |
产品技术规格
Appearance | White to Yellow to Green clear liquid |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...