二乙基二氯磷 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
Diethylphosphoramidous dichloride, (Diethylamino)phosphonous Dichloride
D3244-25G
D3244-5G
TCIAD3244-25GEA
0
CNY
TCIAD3244-25G
TCIAD3244-5G
二乙基二氯磷 ≥98.0% (by GC)
二乙基二氯磷
分子式:
C₄H₁₀Cl₂NP 分子量: 174.00 g/mol 沸点: 189°C 闪点: 83°C 存储条件: Refrigerator |
MDL编号:
MFCD00013614 CAS号: 1069-08-5 UN号: 3265 ADR: 8 II |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...