N,N′-双(3-甲基苯基)-N,N′-二苯基-1,1′-联苯-4,4′-二胺 ≥99.0% (by HPLC, total nitrogen), purified by sublimation

供应商: TCI

Synonyms: TPD

D3236-1G D3236-5G
TCIAD3236-1GEA 0 CNY
TCIAD3236-1G TCIAD3236-5G
N,N′-双(3-甲基苯基)-N,N′-二苯基-1,1′-联苯-4,4′-二胺 ≥99.0% (by HPLC, total nitrogen), purified by sublimation
N,N′-双(3-甲基苯基)-N,N′-二苯基-1,1′-联苯-4,4′-二胺
分子式: C₃₈H₃₂N₂
分子量: 516.6741 g/mol
MDL编号: MFCD00144965
CAS号: 65181-78-4
EINECS: 413-810-8
UN号: 3077
ADR: 9 III

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (with Total Nitrogen) min. 99.0 %
Melting point 168.0 to 172.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR