2,4-Dimethyl-5-phenylthiophene ≥98.0% (by GC)

供应商: TCI

D3228-1G D3228-5G
TCIAD3228-1GEA 0 CNY
TCIAD3228-1G TCIAD3228-5G
2,4-Dimethyl-5-phenylthiophene ≥98.0% (by GC)
2,4-Dimethyl-5-phenylthiophene
分子式: C₁₂H₁₂S
分子量: 188.29 g/mol
沸点: 151°C
存储条件: Refrigerator
CAS号: 57021-49-5

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR