3,3-Dimethyl-1-indanone ≥95.0% (by GC)

供应商: TCI

Synonyms: 3,3-Dimethyl-2,3-dihydro-1H-inden-1-one

D3160-1G D3160-25G D3160-5G
TCIAD3160-1GEA 0 CNY
TCIAD3160-1G TCIAD3160-25G TCIAD3160-5G
3,3-Dimethyl-1-indanone ≥95.0% (by GC)
3,3-Dimethyl-1-indanone
分子式: C₁₁H₁₂O
分子量: 160.22 g/mol
沸点: 122°C
存储条件: Refrigerator
MDL编号: MFCD01846169
CAS号: 26465-81-6

现在订购

产品技术规格

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR