2,3-二甲基蒽醌 ≥95.0% (by GC)
供应商: TCI
|
Warning
|
D2980-1G
D2980-5G
TCIAD2980-1GEA
0
CNY
TCIAD2980-1G
TCIAD2980-5G
2,3-二甲基蒽醌 ≥95.0% (by GC)
2,3-二甲基蒽醌
分子式:
C₁₆H₁₂O₂ 分子量: 236.27 g/mol |
CAS号:
6531-35-7 |
产品技术规格
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 210.0 to 214.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...