Methyl-2,5-dihydrothiophene-3-carboxylate ≥95.0% (by GC)
供应商: TCI
Synonyms: Methyl 2,5-dihydrothiophene-3-carboxylate, 2,5-Dihydrothiophene-3-carboxylic Acid Methyl Ester
D2882-1G D2882-5G
TCIAD2882-1GEA 0 CNY
TCIAD2882-1G TCIAD2882-5G
Methyl-2,5-dihydrothiophene-3-carboxylate ≥95.0% (by GC)
Methyl-2,5-dihydrothiophene-3-carboxylate
分子式: C₆H₈O₂S
分子量: 144.19 g/mol
存储条件: Refrigerator
MDL编号: MFCD04038417
CAS号: 67488-46-4
现在订购
产品技术规格
Appearance White to Orange to Green powder to lump
Purity (GC) min. 95.0 %
Melting point 34.0 to 38.0 deg-C