2,2-二乙氧基丙腈 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2,2-Diethoxypropanenitrile

D2866-25ML D2866-5ML
TCIAD2866-25MLEA 0 CNY
TCIAD2866-25ML TCIAD2866-5ML
2,2-二乙氧基丙腈 ≥98.0% (by GC)
2,2-二乙氧基丙腈
分子式: C₇H₁₃NO₂
沸点: 79°C
闪点: 44°C
MDL编号: MFCD00209573
CAS号: 56011-12-2
UN号: 1993
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR