(+)-2,2′-异亚丙基双[(4R)-4-苯基-2-噁唑啉] ≥96.0% (by GC)

供应商: TCI
Warning

Synonyms: (R,R)-2,2′-异亚丙基双(4-苯基-2-噁唑啉), (R,R)-2,2′-双(4-苯基-2-噁唑啉-2-基)丙烷

D2823-1G D2823-250MG D2823-5G
TCIAD2823-1GEA 0 CNY
TCIAD2823-1G TCIAD2823-250MG TCIAD2823-5G
(+)-2,2′-异亚丙基双[(4R)-4-苯基-2-噁唑啉] ≥96.0% (by GC)
(+)-2,2′-异亚丙基双[(4R)-4-苯基-2-噁唑啉]
分子式: C₂₁H₂₂N₂O₂
存储条件: Refrigerator
MDL编号: MFCD00192244
CAS号: 150529-93-4

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 96.0 %
Specific rotation [a]20/D +155.0 to +165.0 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR