2,2-Diisobutyl-1,3-propanediol ≥98.0% (by GC)

供应商: TCI

Synonyms: 2,2-Dihydroxy-2,2-diisobutylpropane

D2777-25G D2777-5G
TCIAD2777-25GEA 0 CNY
TCIAD2777-25G TCIAD2777-5G
2,2-Diisobutyl-1,3-propanediol ≥98.0% (by GC)
2,2-Diisobutyl-1,3-propanediol
分子式: C₁₁H₂₄O₂
沸点: 160°C
MDL编号: MFCD00986394
CAS号: 10547-96-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR