3,4-二-O-乙酰基-1,5-酐-2,6-双脱氧-L-阿拉伯-己-1-糖醇 ≥95.0% (通过GC)

供应商: TCI

D2752-1G D2752-5G
TCIAD2752-1GEA 0 CNY
TCIAD2752-1G TCIAD2752-5G
3,4-二-O-乙酰基-1,5-酐-2,6-双脱氧-L-阿拉伯-己-1-糖醇 ≥95.0% (通过GC)
3,4-二-O-乙酰基-1,5-酐-2,6-双脱氧-L-阿拉伯-己-1-糖醇
分子式: C₁₀H₁₄O₅
存储条件: Refrigerator
MDL编号: MFCD00074970
CAS号: 34819-86-8
EINECS: 252-228-7

现在订购

产品技术规格

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 95.0 %
Specific rotation [a]20/D +61.0 to +63.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR