3,4-二甲氧基噻吩 ≥98.0% (通过GC)

供应商: TCI

D2728-1G D2728-5G
TCIAD2728-1GEA 0 CNY
TCIAD2728-1G TCIAD2728-5G
3,4-二甲氧基噻吩 ≥98.0% (通过GC)
3,4-二甲氧基噻吩
分子式: C6H8O2S
闪点: 107°C
MDL编号: MFCD01096546
CAS号: 51792-34-8

现在订购

产品技术规格

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR