Dibutyldichlorosilane ≥94.0% (by GC)

供应商: TCI
Danger

D2704-25G D2704-5G
TCIAD2704-25GEA 0 CNY
TCIAD2704-25G TCIAD2704-5G
Dibutyldichlorosilane ≥94.0% (by GC)
Dibutyldichlorosilane
分子式: C₈H₁₈Cl₂Si
沸点: 109°C
闪点: 64°C
MDL编号: MFCD00053202
CAS号: 3449-28-3
UN号: 2987
ADR: 8,II

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR