Diphenyl(methyl)sulphonium tetrafluoroborate ≥97.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: Methyldiphenylsulphonium Tetrafluoroborate, Diphenyl(methyl)sulphonium Tetrafluoroborate

D2685-1G D2685-5G
TCIAD2685-1GEA 0 CNY
TCIAD2685-1G TCIAD2685-5G
Diphenyl(methyl)sulphonium tetrafluoroborate ≥97.0% (by titrimetric analysis)
Diphenyl(methyl)sulphonium tetrafluoroborate
分子式: C₁₃H₁₃BF₄S
MDL编号: MFCD00051861
CAS号: 10504-60-6
EINECS: 234-032-3
UN号: 1759
ADR: 8,III

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 97.0 %
Melting point 64.0 to 68.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR