2,4-Diethyl-1,5-pentanediol (DL- and meso- mixture) ≥93.0% (by GC)

供应商: TCI
Danger

D2619-25ML D2619-500ML
TCIAD2619-25MLEA 0 CNY
TCIAD2619-25ML TCIAD2619-500ML
2,4-Diethyl-1,5-pentanediol (DL- and meso- mixture) ≥93.0% (by GC)
2,4-Diethyl-1,5-pentanediol (DL- and meso- mixture)
分子式: C₉H₂₀O₂
分子量: 160.26 g/mol
沸点: 150°C
闪点: 143°C
MDL编号: MFCD01861285
CAS号: 57987-55-0
EINECS: 429-310-8

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 % (total of isomers)
Specific gravity (20/20) 0.9480 to 0.9520
Refractive index n20/D 1.4590 to 1.4630


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR