2,2'-Dinitrodibenzyl ≥98.0% (通过GC)

供应商: TCI
Warning

D2602-25G D2602-5G
TCIAD2602-25GEA 0 CNY
TCIAD2602-25G TCIAD2602-5G
2,2'-Dinitrodibenzyl ≥98.0% (通过GC)
2,2'-Dinitrodibenzyl
分子式: C₁₄H₁₂N₂O₄
MDL编号: MFCD00024296
CAS号: 16968-19-7
EINECS: 241-043-7

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 123.0 deg-C
Solubility in Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR