Diphenylphosphinic anhydride ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
D2484-1G
D2484-5G
TCIAD2484-1GEA
0
CNY
TCIAD2484-1G
TCIAD2484-5G
Diphenylphosphinic anhydride ≥98.0% (by titrimetric analysis)
Diphenylphosphinic anhydride
分子式:
C₂₄H₂₀O₃P₂ |
MDL编号:
MFCD01112176 CAS号: 5849-36-5 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Neutralization back titration) | min. 98.0 % |
Melting point | 141.0 to 146.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...