2-甲基-3-二甲氨基-2-丙烯醛 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
alpha-Methyl-beta-dimethylaminoacrolein
D2483-25G
D2483-5G
TCIAD2483-25GEA
0
CNY
TCIAD2483-25G
TCIAD2483-5G
2-甲基-3-二甲氨基-2-丙烯醛 ≥97.0% (by GC)
2-甲基-3-二甲氨基-2-丙烯醛
分子式:
C₆H₁₁NO 分子量: 113.16 g/mol |
MDL编号:
MFCD00075471 CAS号: 19125-76-9 |
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...