(R,R)-(-)-N,N′-二甲基-1,2-环己二胺 ≥94.0% (by GC)

供应商: TCI
Danger

Synonyms: (R,R)-(-)-N,N′-二甲基-1,2-二氨基环己烷, 反式-1,2-双(甲氨基)环己烷

D2459-1G D2459-100MG
TCIAD2459-1GEA 0 CNY
TCIAD2459-1G TCIAD2459-100MG
(R,R)-(-)-N,N′-二甲基-1,2-环己二胺 ≥94.0% (by GC)
(R,R)-(-)-N,N′-二甲基-1,2-环己二胺
分子式: C₈H₁₈N₂
存储条件: Refrigerator
MDL编号: MFCD00671527
CAS号: 68737-65-5
UN号: 3259
ADR: 8,III

现在订购

产品技术规格

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 94.0 %
Optical purity (GC) min. 98.0 ee%
Melting point 43.0 to 47.0 deg-C
Specific rotation [a]20/D -138 to -148 deg (C=4, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR