cis-13-Docosenol ≥98.0% (by GC)

供应商: TCI

Synonyms: Erucyl Alcohol, Cis-13-docosenol

D2174-1G D2174-5G
TCIAD2174-1GEA 0 CNY
TCIAD2174-1G TCIAD2174-5G
cis-13-Docosenol ≥98.0% (by GC)
cis-13-Docosenol
分子式: C₂₂H₄₄O
分子量: 324.59 g/mol
沸点: 225°C
MDL编号: MFCD00055936
CAS号: 629-98-1
EINECS: 211-122-0

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Freezing point 31.0 to 34.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR