Dimethylthiophosphinoyl chloride ≥97.0% (通过GC)
供应商: TCI
|
Danger
|
D2159-1G
D2159-5G
TCIAD2159-1GEA
0
CNY
TCIAD2159-1G
TCIAD2159-5G
Dimethylthiophosphinoyl chloride ≥97.0% (通过GC)
Dimethylthiophosphinoyl chloride
分子式:
C₂H₆ClPS 沸点: 78°C 存储条件: Refrigerator |
MDL编号:
MFCD00014454 CAS号: 993-12-4 EINECS: 213-606-7 UN号: 3265 ADR: 8,II |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...