1,3-Diiminoisoindoline ≥98.0% (通过滴定分析)

供应商: TCI
Warning

D2079-25G D2079-5G
TCIAD2079-25GEA 0 CNY
TCIAD2079-25G TCIAD2079-5G
1,3-Diiminoisoindoline ≥98.0% (通过滴定分析)
1,3-Diiminoisoindoline
分子式: C₈H₇N₃
MDL编号: MFCD00010062
CAS号: 57500-34-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR