3,4-Diacetoxybenzaldehyde ≥98.0% (通过GC)

供应商: TCI
Warning

D2042-25G D2042-5G
TCIAD2042-25GEA 0 CNY
TCIAD2042-25G TCIAD2042-5G
3,4-Diacetoxybenzaldehyde ≥98.0% (通过GC)
3,4-Diacetoxybenzaldehyde
分子式: C₁₁H₁₀O₅
MDL编号: MFCD00143057
CAS号: 67727-64-4

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR