2,2-Di(2-tetrahydrofuryl)propane ≥96.0% (通过GC)

供应商: TCI
Warning

D2034-25ML D2034-500ML
TCIAD2034-25MLEA 0 CNY
TCIAD2034-25ML TCIAD2034-500ML
2,2-Di(2-tetrahydrofuryl)propane ≥96.0% (通过GC)
2,2-Di(2-tetrahydrofuryl)propane
分子式: C₁₁H₂₀O₂
沸点: 146°C
闪点: 95°C
MDL编号: MFCD00142763
CAS号: 89686-69-1

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR