1,3-Dimethyl-5-pyrazolone ≥98.0% (by HPLC, titration analysis)
供应商: TCI
Synonyms:
2,4-Dihydro-2,5-dimethyl-3H-pyrazol-3-one, 1,3-Dimethyl-1H-pyrazol-5(4H)-one
D2031-25G
D2031-500G
TCIAD2031-25GEA
0
CNY
TCIAD2031-25G
TCIAD2031-500G
1,3-Dimethyl-5-pyrazolone ≥98.0% (by HPLC, titration analysis)
1,3-Dimethyl-5-pyrazolone
分子式:
C₅H₈N₂O 分子量: 112.13 g/mol |
MDL编号:
MFCD00067008 CAS号: 2749-59-9 EINECS: 220-389-2 |
产品技术规格
Appearance | White to Light red to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 116.0 to 121.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...