二巯基马来腈 ≥90.0% (by total nitrogen basis)

供应商: TCI
Danger

Synonyms: Dimercaptomaleonitrile Disodium Salt

D1930-1G D1930-5G
TCIAD1930-1GEA 0 CNY
TCIAD1930-1G TCIAD1930-5G
二巯基马来腈 ≥90.0% (by total nitrogen basis)
二巯基马来腈
分子式: C₄H₂N₂S₂
分子量: 186.16 g/mol
MDL编号: MFCD00060122
CAS号: 5466-54-6
UN号: 3439
ADR: 6.1 III

现在订购

产品技术规格

Appearance White to Amber powder to crystal
Purity (with Total Nitrogen) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR