4,6-Diaminoresorcinol dihydrochloride ≥98.0% (by total nitrogen and titration analysis)

供应商: TCI
Warning

Synonyms: 4,6-Dihydroxy-1,3-phenylenediamine Dihydrochloride, 4,6-Diaminobenzene-1,3-diol dihydrochloride

D1888-25G D1888-5G
TCIAD1888-25GEA 0 CNY
TCIAD1888-25G TCIAD1888-5G
4,6-Diaminoresorcinol dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
4,6-Diaminoresorcinol dihydrochloride
分子式: C₆H₈N₂O₂
分子量: 213.06 g/mol
MDL编号: MFCD00143239
CAS号: 16523-31-2

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR