3-(Diethylamino)pyrrolidine ≥98.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: N,N-Diethylpyrrolidin-3-amine

D1857-25G D1857-5G
TCIAD1857-25GEA 0 CNY
TCIAD1857-25G TCIAD1857-5G
3-(Diethylamino)pyrrolidine ≥98.0% (by titrimetric analysis)
3-(Diethylamino)pyrrolidine
分子式: C₈H₁₈N₂
沸点: 84°C
MDL编号: MFCD02184351
CAS号: 20984-81-0
UN号: 2735
ADR: 8,III

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR