4,6-Dimethyl-2-hydroxypyrimidine Hydrochloride ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
2-Hydroxy-4,6-dimethylpyrimidine Hydrochloride, 4,6-Dimethyl-2-pyrimidinol Hydrochloride, 4,6-Dimethylpyrimidin-2-ol hydrochloride
D1823-25G
D1823-250G
TCIAD1823-25GEA
0
CNY
TCIAD1823-25G
TCIAD1823-250G
4,6-Dimethyl-2-hydroxypyrimidine Hydrochloride ≥98.0% (by HPLC, titration analysis)
4,6-Dimethyl-2-hydroxypyrimidine hydrochloride
分子式:
C₆H₈N₂O 分子量: 160.60 g/mol 溶点: 281°C |
MDL编号:
MFCD00035312 CAS号: 34289-60-6 EINECS: 251-919-0 |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...