甲基三甲基硅基二甲基乙烯酮缩二乙醇 ≥95.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
(1-甲氧基-2-甲基-1-烯丙氧基)三甲基硅烷, 1-甲氧基-2-甲基-1-(三甲基硅氧基)丙烯, MTDA
D1789-25ML
D1789-5ML
TCIAD1789-25MLEA
0
CNY
TCIAD1789-25ML
TCIAD1789-5ML
甲基三甲基硅基二甲基乙烯酮缩二乙醇 ≥95.0% (by GC)
甲基三甲基硅基二甲基乙烯酮缩二乙醇
分子式:
C₈H₁₈O₂Si 分子量: 174.32 g/mol 沸点: 52°C 闪点: 30°C |
MDL编号:
MFCD00010232 CAS号: 31469-15-5 UN号: 1993 ADR: 3 III |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...