3-(Diethylamino)propionic acid hydrochloride ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: N,N-Diethyl-beta-alanine Hydrochloride

D1713-25G D1713-5G
TCIAD1713-25GEA 0 CNY
TCIAD1713-25G TCIAD1713-5G
3-(Diethylamino)propionic acid hydrochloride ≥98.0% (by titrimetric analysis)
3-(Diethylamino)propionic acid hydrochloride
分子式: C₇H₁₅NO₂
分子量: 181.66 g/mol
MDL编号: MFCD00012611
CAS号: 15674-67-6
EINECS: 239-751-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 144.0 to 147.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR