蒽-9,10-二腈 ≥98.0% (by HPLC, total nitrogen)

供应商: TCI
Danger

Synonyms: 9,10-二氰基蒽

D1656-1G D1656-5G
TCIAD1656-1GEA 0 CNY
TCIAD1656-1G TCIAD1656-5G
蒽-9,10-二腈 ≥98.0% (by HPLC, total nitrogen)
蒽-9,10-二腈
分子式: C₁₆H₈N₂
分子量: 228.25 g/mol
MDL编号: MFCD00041733
CAS号: 1217-45-4
UN号: 3439
ADR: 6.1 III

现在订购

产品技术规格

Appearance Light yellow to Yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR