N,N-Dimethyl-n-octadecylamine ≥90.0% (by GC)

供应商: TCI
Danger

Synonyms: N,N-Dimethyloctadecan-1-amine, Dymanthine, N,N-Dimethylstearylamine

D1609-25G D1609-250G
TCIAD1609-25GEA 0 CNY
TCIAD1609-25G TCIAD1609-250G
N,N-Dimethyl-n-octadecylamine ≥90.0% (by GC)
N,N-Dimethyl-n-octadecylamine
分子式: C₂₀H₄₃N
分子量: 297.57 g/mol
MDL编号: MFCD00048496
CAS号: 124-28-7
EINECS: 204-694-8
UN号: 3259
ADR: 8 III

现在订购

产品技术规格

Appearance White to Almost white powder to lump
Purity (GC) min. 90.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR