1-(Dimethylisopropylsilyl)imidazole ≥98.0% (通过滴定分析) dimethyl-isopropyl-silylating agent

供应商: TCI
Warning

D1596-1G D1596-5G
TCIAD1596-1GEA 0 CNY
TCIAD1596-1G TCIAD1596-5G
1-(Dimethylisopropylsilyl)imidazole ≥98.0% (通过滴定分析) dimethyl-isopropyl-silylating agent
1-(Dimethylisopropylsilyl)imidazole
分子式: C₈H₁₆N₂Si
存储条件: Refrigerator
MDL编号: MFCD00059891
CAS号: 81452-04-2

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (Nonaqueous Titration) min. 98.0 %
Effect test of GC derivalization Effective as derivatization agent Estradiol, Cholesterol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR