2-甲基-2-丙烯酸-2-(2-甲氧基乙氧基)乙酯 ≥97.0% (by GC) stabilised
供应商: TCI
|
Warning
|
Synonyms:
2-(2-甲氧基乙氧基)甲基丙烯酸乙酯
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
D1531-25ML
D1531-500ML
TCIAD1531-25MLEA
0
CNY
TCIAD1531-25ML
TCIAD1531-500ML
2-甲基-2-丙烯酸-2-(2-甲氧基乙氧基)乙酯 ≥97.0% (by GC) stabilised
2-甲基-2-丙烯酸-2-(2-甲氧基乙氧基)乙酯
分子式:
C₉H₁₆O₄ 分子量: 188.22 g/mol 闪点: 111°C 存储条件: Refrigerator |
MDL编号:
MFCD00059330 CAS号: 45103-58-0 EINECS: 256-190-2 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...