Dansyl-L-isoleucine

供应商: TCI

Synonyms: Dns-Ile-OH, 5-Dimethylaminonaphthalene-1-sulphonyl-L-isoleucine

D1497-1G D1497-100MG
TCIAD1497-1GEA 0 CNY
TCIAD1497-1G TCIAD1497-100MG
Dansyl-L-isoleucine
Dansyl-L-isoleucine
分子式: C₁₈H₂₄N₂O₄S
MDL编号: MFCD00069525
CAS号: 1100-21-6

现在订购

产品技术规格

Purity (HPLC) min. 94.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR