2,2-Dimethyltetrahydrothiazole ≥98.0% (通过GC)

供应商: TCI
Warning

D1491-25ML D1491-5ML
TCIAD1491-25MLEA 0 CNY
TCIAD1491-25ML TCIAD1491-5ML
2,2-Dimethyltetrahydrothiazole ≥98.0% (通过GC)
2,2-Dimethyltetrahydrothiazole
分子式: C₅H₁₁NS
沸点: 61°C
闪点: 55°C
MDL编号: MFCD00014488
CAS号: 19351-18-9
EINECS: 242-980-4
UN号: 1993
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR