Sodium-2,4-dichlorophenoxyacetate monohydrate ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: Na-2,4-D, 2,4-Dichlorophenoxyacetic Acid Sodium Salt

D1319-25G D1319-500G
TCIAD1319-25GEA 0 CNY
TCIAD1319-25G TCIAD1319-500G
Sodium-2,4-dichlorophenoxyacetate monohydrate ≥98.0% (by HPLC, titration analysis)
Sodium-2,4-dichlorophenoxyacetate monohydrate
分子式: C₈H₆Cl₂O₃
MDL编号: MFCD00068284
CAS号: 2702-72-9
EINECS: 220-290-4

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water 6.0 to 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR