Decamethonium iodide

供应商: TCI
Danger

D1315-25G D1315-5G
TCIAD1315-25GEA 0 CNY
TCIAD1315-25G TCIAD1315-5G
Decamethonium iodide
Decamethonium iodide
分子式: C₁₆H₃₈I₂N₂
MDL编号: MFCD00031553
CAS号: 1420-40-2
EINECS: 215-816-4
UN号: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2e218397a2396cd2e232aa7bbb326ed644b68f8&vid=a2e218397a2396cd2e232aa7bbb326ed644b68f8&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR