三氟乙酸钠 97%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
三氟乙酸
157930250
157931000
ACRO157930250EA
0
CNY
ACRO157930250
ACRO157931000
三氟乙酸钠 97%
三氟乙酸钠
分子式:
C₂F₃NaO₂ 分子量: 136.01 g/mol 溶点: 207…209 °C |
MDL编号:
MFCD00013217 CAS号: 2923-18-4 EINECS: 220-879-6 |
产品技术规格
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 205 to 207 °C |
Titration with HClO4 | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...