2,4-二羟基-5,6-二甲基嘧啶 ≥99.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 5,6-二甲基尿嘧啶

D1136-1G D1136-5G
TCIAD1136-1GEA 0 CNY
TCIAD1136-1G TCIAD1136-5G
2,4-二羟基-5,6-二甲基嘧啶 ≥99.0% (by HPLC, titration analysis)
2,4-二羟基-5,6-二甲基嘧啶
分子式: C₆H₈N₂O₂
MDL编号: MFCD00010708
CAS号: 26305-13-5
EINECS: 247-601-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR