4,4'-Di-n-octyloxyazoxybenzene ≥97.0% (通过HPLC,总氮)
供应商: TCI
D1123-1G
D1123-5G
TCIAD1123-1GEA
0
CNY
TCIAD1123-1G
TCIAD1123-5G
4,4'-Di-n-octyloxyazoxybenzene ≥97.0% (通过HPLC,总氮)
4,4'-Di-n-octyloxyazoxybenzene
分子式:
C₂₈H₄₂N₂O₃ |
MDL编号:
MFCD00059445 CAS号: 25729-12-8 |
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Mesomorphic range | 79.0 to 112.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...