十二烷基苯磺酸 mixture ≥95.0% (by titrimetric analysis), soft type
供应商: TCI
|
Danger
|
Synonyms:
Alkylbenzenesulphonic acid, Laurylbenzenesulphonic acid
D0989-25G
D0989-500G
TCIAD0989-25GEA
0
CNY
TCIAD0989-25G
TCIAD0989-500G
十二烷基苯磺酸 mixture ≥95.0% (by titrimetric analysis), soft type
十二烷基苯磺酸
分子式:
C₁₈H₃₀O₃S 分子量: 326.5 沸点: 315 溶点: 10 密度: 0.9 闪点: 211°C |
MDL编号:
MFCD00066677 CAS号: 68584-22-5 EINECS: 271-528-9 UN号: 2586 ADR: 8 III |
产品技术规格
Appearance | Orange to Brown to Dark red clear liquid |
Purity (Phase splitting method) | min. 95.0 % |
Neutralization Value | 177.0 to 186.0 |
Petroleum ether soluble matter | max. 2.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...