2,4-Dimethyl-1,3-dioxane ≥98.0% (通过GC)
供应商: TCI
|
Danger
|
D0708-10ML
D0708-25ML
TCIAD0708-10MLEA
0
CNY
TCIAD0708-10ML
TCIAD0708-25ML
2,4-Dimethyl-1,3-dioxane ≥98.0% (通过GC)
2,4-Dimethyl-1,3-dioxane
分子式:
C₆H₁₂O₂ 沸点: 118°C |
MDL编号:
MFCD00023821 CAS号: 766-20-1 EINECS: 212-162-1 UN号: 2707 ADR: 3,II |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...