DL-Glyceric acid 20% ∼2 mol/L in water

供应商: TCI
Warning

Synonyms: DL-2,3-Dihydroxypropionic acid, 2,3-Dihydroxypropanoic acid

D0602-100G D0602-25G
TCIAD0602-100GEA 0 CNY
TCIAD0602-100G TCIAD0602-25G
DL-Glyceric acid 20% ∼2 mol/L in water
DL-Glyceric acid
分子式: C₃H₆O₄
分子量: 106.08 g/mol
MDL编号: MFCD00065927
CAS号: 473-81-4
EINECS: 207-472-9

现在订购

产品技术规格

Appearance Colorless to Red to Green clear liquid
Concentration (Neutralization titration) 20.0 to 22.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR