1,4-二氢-2,4,6-三甲基-3,5-吡啶二甲酸二乙酯 ≥98.0% (by GC)

供应商: TCI

Synonyms: DDC, 3,5-二乙氧基羰基-1,4-二氢-2,4,6-三甲基吡啶

D0558-1G D0558-5G
TCIAD0558-1GEA 0 CNY
TCIAD0558-1G TCIAD0558-5G
1,4-二氢-2,4,6-三甲基-3,5-吡啶二甲酸二乙酯 ≥98.0% (by GC)
1,4-二氢-2,4,6-三甲基-3,5-吡啶二甲酸二乙酯
分子式: C₁₄H₂₁NO₄
分子量: 267.33 g/mol
MDL编号: MFCD00005950
CAS号: 632-93-9
EINECS: 211-188-0

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 129.0 to 133.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR