Bromopyrogallol red

供应商: TCI

D0207-1G D0207-5G
TCIAD0207-1GEA 0 CNY
TCIAD0207-1G TCIAD0207-5G
Bromopyrogallol red
Bromopyrogallol red
分子式: C₁₉H₁₀Br₂O₈S
MDL编号: MFCD00148865
CAS号: 16574-43-9
EINECS: 240-632-6

现在订购

产品技术规格

Appearance Red to Dark blue to Black powder to crystal
Lambda max. 550.0 to 560.0 nm (pH 6.0 buffer sol.)
Molar extinction coefficient min. 35,000 (pH 6.0 buffer sol., 550.0 to 560.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR