2,6-二氨基庚二酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 2,6-二氨基蒲桃酸

D0112-1G D0112-5G
TCIAD0112-1GEA 0 CNY
TCIAD0112-1G TCIAD0112-5G
2,6-二氨基庚二酸 ≥98.0% (by titrimetric analysis)
2,6-二氨基庚二酸
分子式: C₇H₁₄N₂O₄
分子量: 190.20 g/mol
MDL编号: MFCD00002637
CAS号: 583-93-7
EINECS: 209-524-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in 1mol/L HCl almost transparency
Water max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR