去氢胆酸 ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: 3,7,12-三氧-5β-胆烷酸

D0042-25G D0042-250G
TCIAD0042-25GEA 0 CNY
TCIAD0042-25G TCIAD0042-250G
去氢胆酸 ≥98.0% (by titrimetric analysis)
去氢胆酸
分子式: C₂₄H₃₄O₅
MDL编号: MFCD00066410
CAS号: 81-23-2
EINECS: 201-335-7
Merck Index: 14,02868

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 236.0 to 240.0 deg-C
Specific rotation [a]20/D +29.0 to +32.0 deg (C=2, Dioxane)
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR