L-Carnitine-L-tartrate ≥98.0% (by titrimetric analysis)
供应商: TCI
Synonyms:
Bis[[(R)-3-carboxy-2-hydroxypropyl]trimethylammonium] L-Tartrate
C2625-25G
C2625-5G
TCIAC2625-25GEA
0
CNY
TCIAC2625-25G
TCIAC2625-5G
L-Carnitine-L-tartrate ≥98.0% (by titrimetric analysis)
L-Carnitine-L-tartrate
分子式:
C₇H₁₅NO₃ 分子量: 236.24 g/mol |
MDL编号:
MFCD04974128 CAS号: 36687-82-8 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...